Learn More
Sigma Aldrich Fine Chemicals Biosciences Transfer Pipette capacity
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS206371C500EA
Encompass
The Transfer Pipette is a one-piece unbreakable polyethylene plastic pipette and is well-suited for blood banking urinalysis hematology wet chemistry microbiology and serology.
Catalog No. 50-290-0322