Learn More
Cole-Parmer TRANSFER PIPETTE,FINE-TIP,500/
Supplier:  Cole-Parmer 0622667
Encompass_Preferred
Transfer Pipette, Fine-tip, 5.8 mL; 500/Pk.Easily draw and deliver precise solutions Deliver repeatable, uniform dropsEliminate cross contamination with these low cost, disposable LDPE pipettes
Catalog No. NC1929333