Learn More
Sigma Aldrich Fine Chemicals Biosciences Transfer pipette polyethyl
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z135070500EA
Encompass
Disposable transfer pipettes are a great way to transfer liquids without the worry of cross-contamination. Available in 19 styles our extensive range is sure to have one that fits your needs. Available as non-sterile sterile or sterile and wrapped in packs of 10.Unified design with built-in pipette bulbSafe convenient use and disposalReduces loss of cells and proteins that bind to glassHeat sealable to create liquid-tight storageNon-autoclavable
Catalog No. 50-290-1578