Learn More
Thomas Scientific Transfer Pipet, 5.0mL, Large Bulb, Graduated to 1mL, 145mm, 500/Dispenser Box
SDP
Catalog No. NC0221960
Encompass
Material: Low Density Polyethylene (LDPE) Packaging: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean. Each dispenser box contains 500 pipets.
Catalog No. NC0221960 Supplier Thomas Scientific Supplier No. 1149K67
Material: Low Density Polyethylene (LDPE) Packaging: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean. Each dispenser box contains 500 pipets.