Learn More
Chemplex Industries TRANSFER PIPETTES 8.7ML
Catalog No. 50249537
Encompass_Preferred
CHEMPLEX TRANSFER PIPETTES, 8.7 Overall Capacity, mL, Fine tip; large bulb
Catalog No. 50-249-537 Supplier Chemplex Industries Supplier No. 800234
CHEMPLEX TRANSFER PIPETTES, 8.7 Overall Capacity, mL, Fine tip; large bulb