Learn More
TWD Scientific LLC Transfer Pipette, Twdbiox, 1Ml, Disposable, Sterile, Polyethylene, Molded Graduations, Individually Wrapped, 2000/Case.
SDP
Catalog No. 502285027 Shop All TWD Scientific LLC Products
Encompass_Preferred
1Ml Disposable Polyethylene Sterile General Purpose Transfer Pippets (Length: 160Mm | Capacity: 6.2Ml). Pippets Have Molded Graduations At Each 0.25Ml Up To 1Ml (26 Drops/Ml). Pippets Come Individually Wrapped. 500 Pipettes/Pack, 4 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5027 Supplier TWD Scientific LLC Supplier No. BX300135A11
1Ml Disposable Polyethylene Sterile General Purpose Transfer Pippets (Length: 160Mm | Capacity: 6.2Ml). Pippets Have Molded Graduations At Each 0.25Ml Up To 1Ml (26 Drops/Ml). Pippets Come Individually Wrapped. 500 Pipettes/Pack, 4 Packs/Case, Price/Case