Learn More
Grainger Transfer Pump Pressure Booster 1/2 HP
Catalog No. 502751151
Encompass_Preferred
The Grainger Approved Transfer Pump Pressure Booster is a 1/2 HP electric pump designed for transferring non-flammable liquids. It has a thermoplastic body and impeller, can pump up to 20 GPM at 50 PSI, and has a maximum pressure of 100 PSI. The pump has a 1/2" NPT inlet and outlet, operates on 115V power, and is CSA certified. It's ideal for transferring water, chemicals, petroleum products and more. The compact size allows it to fit in tight spaces.
Catalog No. 50-275-1151 Supplier Grainger Supplier No. 157F69
The Grainger Approved Transfer Pump Pressure Booster is a 1/2 HP electric pump designed for transferring non-flammable liquids. It has a thermoplastic body and impeller, can pump up to 20 GPM at 50 PSI, and has a maximum pressure of 100 PSI. The pump has a 1/2" NPT inlet and outlet, operates on 115V power, and is CSA certified. It's ideal for transferring water, chemicals, petroleum products and more. The compact size allows it to fit in tight spaces.