Learn More
Description
- One unit will yield an A260 of 1.0 in 1.0 mL water (1 cm light path).
Specifications
Specifications
| Format | Vial (powder) |
| For Use With (Application) | DNA and RNA Purification, Enzymes, Substrates, Inhibitors |
| Product Type | Transfer Ribonucleic Acid |
| Quantity | 5 kU |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.