Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transfer Ribonucleic Acid, from Baker's Yeast, MP Biomedicals™

Catalog No. ICN15653490 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kU
500 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15653490 500 U
02-156-53494 5 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15653490 Supplier MP Biomedicals, Inc Supplier No. 0215653490
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Transfer Ribonucleic Acid can be used for a tRNA standard and in reactions involving tRNAs, such as assays for tRNA synthetase activity.

  • One unit will yield an A260 of 1.0 in 1.0 mL water (1 cm light path).
TRUSTED_SUSTAINABILITY

Specifications

Format Vial (powder)
Product Type Transfer Ribonucleic Acid
Quantity 500 U

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.