Learn More
Genscript Corporation eBlot L1 Transfer sandwich without membrane (with buffers for NC)
Catalog No. NC1677871
Encompass
eBlot L1 is a highly efficient wet protein transfer system, which uses a patented technology developed by GenScript. eBlot L1 combines the high stability and efficiency of the traditional wet transfer with the rapidness and convenience of the semi-dry transfer system. The device allows the efficient transfer of small, medium as well as large molecular weight proteins within 15 minutes.
Each kit contains 2x 1L eBlot L1 NC Membrane Transfer Buffer (5X), 2x 15mL eBlot L1 NC Membrane Equilibration Buffer (10X) and 2x 15pk eBlot L1 Transfer Sponge. The kit is for 30x transfers with standard program.
Catalog No. NC1677871 Supplier Genscript Corporation Supplier No. L00724
eBlot L1 is a highly efficient wet protein transfer system, which uses a patented technology developed by GenScript. eBlot L1 combines the high stability and efficiency of the traditional wet transfer with the rapidness and convenience of the semi-dry transfer system. The device allows the efficient transfer of small, medium as well as large molecular weight proteins within 15 minutes.
Each kit contains 2x 1L eBlot L1 NC Membrane Transfer Buffer (5X), 2x 15mL eBlot L1 NC Membrane Equilibration Buffer (10X) and 2x 15pk eBlot L1 Transfer Sponge. The kit is for 30x transfers with standard program.