Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Transfer Tape

Catalog No. 19108028 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Translucent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-108-028 Translucent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-108-028 Supplier Brady™ Supplier No. 76737
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Light but hi-tack adhesive solution that allows to transfer shapes or text, printed from BBP37/Globalmark printer, directly from liner to desired final surface - without losing spacing or alignment. Brady™ Transfer Tape is used with GlobalMark™ Label Maker; BBP™37 Label Maker.

  • Users cut and print shapes or pre-spaced text from standard adhesive tape with their BBP37/Globalmark printer
  • Excess liner is removed leaving only the cut-out shapes and text
  • Users apply and press BBP37/Globalmark printer transfer tape over the cut-outs in order to maintain alignment among all components
  • Tape is carefully lifted, pulling the cut-outs away from liner in order to keep shapes and text grouped together, while still exposing the adhesive side for the final transfer to a another surface, such as a window, wall, door or blank rigid sign panel
  • Position and align to desired final surface

Specifications

Product Type Transfer Tape
Color Translucent
For Use With (Equipment) GlobalMark Label Maker, BBP37 Color and Cut Sign and Label Printer
Dimensions (L x W) 300 ft. x 4.5 in.
Adhesive Type Removable
Finish Matte
Format Roll
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.