Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Transferpette Electronic Stand

Catalog No. 14381080 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Number of Channels:
1 or 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
14-381-080 1 or 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-381-080 Supplier BRANDTECH™ BRAND™ Supplier No. 705384
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with the Transferpette electronic

  • For use with Transferpette electronic
  • Accessory
  • One or eight channels
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Assorted
Disposable Reusable
For Use With BRAND Transferpette Electronic
Material Polypropylene
Model Pipette Stand
Number of Channels 1 or 8
Sterility Non-sterile
Type Pipette Holder
For Use With (Equipment) Transferpette Electronic Stand
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.