Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ Transferpette™ pro Wall Mounts

Catalog No. 13200195 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Material:
Polycarbonate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
13-200-195 Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-200-195 Supplier BRANDTECH™ Supplier No. 704882
Add to Cart
Add to Cart

Spare part for Transferpette™ pro

  • Spare part
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color Light Gray
Holds Manual Pipette
Material Polycarbonate
Positions 1
Shape Horizontal
Stackable No
Sterility Non-sterile
For Use With (Equipment) Transferpette pro
Product Type Pipette Wall Mount
Shelves Removable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.