Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Transferpette™ S Pipette Bench Rack

Catalog No. 14381059 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Holds:
Pipettes (6)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
14-381-059 Pipettes (6)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-381-059 Supplier BRANDTECH™ BRAND™ Supplier No. 704807
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for use with Transferpette™ S single and multichannel pipettes.

  • Holds six instruments
  • Rotates for easy pipette access
  • Pipettes sold separately
TRUSTED_SUSTAINABILITY

Specifications

Description Rotates
Autoclavable Not Autoclavable
Color White
Holds Pipettes (6)
Material Plastic
Positions 6
Shape Circular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Transferpette S Pipettes
Product Type Benchtop Rack
Shelves None
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.