Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Transferrin (Apo), bovine plasma, 98+%

Catalog No. AAJ61626ME
Change view
Click to view available options
Quantity:
100mg
500mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61626ME 500mg
AAJ61626MC 100mg
AAJ6162603 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61626ME Supplier Thermo Scientific Chemicals Supplier No. J61626ME
Add to Cart

missing translation for 'validMainframeMsg'

For research use

Apo-Transferrin is also an important component used for the cultivation of mammalian cells in-vitro because it acts as a transport factor for defined culture media. This property is crucial for long-term mammalian cell growth in-vitro since ATF operates in media by binding to contaminating metal ions. Some other common uses for Apo-Transferrin are affinity purification and receptor mediated transfection of DNA-like molecules into cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture - Iron Delivery
Formulation Lyophilized powder
Quantity 500mg
Source Bovine plasma
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Transferrin (Apo), Bovine Plasma, ≥98%
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.