Learn More
Athens Research And Technology Inc TRANSFERRIN APO LW ENDTX 100MG
Supplier:  Athens Research And Technology Inc 1616A32001LEL100MG
Encompass_Preferred
Apotransferrin, Human Plasma, LOW ENDOTOXIN, Suitable for Cell Culture Use: 100 MG; Lyophilized from sterile 10 mM phosphate buffer, pH 7.4. Endotoxin Level: <=0.1EU/mg; MW: 80,000. Purity: > =95% by SDS-PAGE; Iron:<30 ug/gm.
Catalog No. 50-489-663