Learn More
Athens Research And Technology Inc TRANSFERRIN CELL CULTURE HU 1G
Supplier:  Athens Research And Technology Inc 1616032001LEL1GR
Encompass_Preferred
Transferrin, Human Plasma, Low Endotoxin Level: 1 GRAM; Lyophilized from 20 mM Na phosphate, pH 7.4, with 150 mM NaCl.; MW 80,000; Endotoxin Level: <=0.1EU/mg; Iron Content: > = 1100UG/GR; Purity: > =95% by SDS-PAGE. Native; Purified from human plasma in the Athens Research & Technology laboratory.
Catalog No. 50-489-925