Learn More
Enzo Life Sciences Transferrin ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1430001
Encompass_Preferred
The Transferrin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours. Assay Time: 2 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human Dog.
Catalog No. 50-201-3837