Learn More
Genscript Corporation Transferrin His Bovine
Catalog No. 5000057503
Encompass
Transferrin is a blood plasma glycoprotein that plays a central role in iron metabolism and is responsible for ferric-ion delivery Transferrin functions as the most critical ferric pool in the body It transports iron through the blood to various tissues such as the liver spleen and bone marrow
Catalog No. 50-000-57503 Supplier Genscript Corporation Supplier No. Z03736100
Transferrin is a blood plasma glycoprotein that plays a central role in iron metabolism and is responsible for ferric-ion delivery Transferrin functions as the most critical ferric pool in the body It transports iron through the blood to various tissues such as the liver spleen and bone marrow