Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Transferrin (Holo), bovine plasma, 98+%

Catalog No. AAJ6104603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6104603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6104603 Supplier Thermo Scientific Chemicals Supplier No. J6104603
Add to Cart

missing translation for 'validMainframeMsg'

 

Transferrin (Holo) is the primary iron binding and cell delivery molecule present in serum. Serum-free cell culture systems require a delivery format for iron. It is the preferential delivery form of iron because cells process transferrin bound iron in a physiologically appropriate way through transferrin receptors on the cell surface. It can be used with a wide range of cells types. It is loaded with iron prior to use and can be added directly to iron poor or iron-free cell culture media. It has been used for study in treatment of breast cancer cells with holo-transferrin in combination with dihydroartemisinin results in increased tumor cell death due to elevated levels of intracellular ferrous iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture - Iron Delivery
Formulation Lyophilized powder
Quantity 1 g
Source Bovine plasma
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Transferrin (Holo), Bovine Plasma, ≥98%
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.