Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Transferrin ELISA Kit

Catalog No. EHTF
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHTF 96 Tests
EHTFX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHTF Supplier Invitrogen™ Supplier No. EHTF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Transferrin ELISA quantitates Hu Transferrin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Transferrin. Principle of the method The Human Transferrin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis.Serum transferrin may also have a further role in stimulating cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02787
Assay Range 1.029-750 ng/mL
Assay Sensitivity 1 ng/mL
Calibration Range 1 ng/mL to 750 ng/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human Transferrin. Other species not determined.
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI266983; beta-1 metal-binding globulin; Cd176; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; HEL-S-71p; HP; hpx; hypotransferrinemia with hemochromatosis; ICA; Inhibitor of carbonic anhydrase; lactoferrin; liver regeneration-related protein LRRG03; liver transferrin; LOC100146633; PICA; porcine inhibitor of carbonic anhydrase; porcine inhibitor of carbonic anhydrase precursor; PRO1400; PRO1557; PRO2086; Serotransferrin; siderophilin; TF; Tfn; TFQTL1; transferin; Transferrin; transferrin precursor; TrfAI266983; beta-1 metal-binding globulin; Cd176; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; HEL-S-71p; HP; hpx; hypotransferrinemia with hemochromatosis; ICA; Inhibitor of carbonic anhydrase; lactoferrin; liver regeneration-related protein LRRG03; liver transferrin; LOC100146633; PICA; porcine inhibitor of carbonic anhydrase; porcine inhibitor of carbonic anhydrase precursor; PRO1400; PRO1557; PRO2086; Serotransferrin; siderophilin; TF; Tfn; TFQTL1; transferin; Transferrin; transferrin precursor; Trf; HEL-S-71p; PRO1557; PRO2086QTL1; TF
Gene ID (Entrez) 7018
Gene Symbol Tf
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Transferrin, Beta-1 metal-binding globulin, Siderophilin
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Cardiovascular Research
Sample Volume Plasma 2 μL, Serum 2 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym PRO1400, PRO1557, PRO2086QTL1, TF
Target Transferrin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.