Learn More
Enzo Life Sciences Transferrin polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIVAPEN004D
Encompass_Preferred
Host: Rabbit. Immunogen: Human Transferrin. UniProt ID: P02787. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Guinea pig, Dog. Applications: IP, WB. Recommended Dilutions: Western Blot (1:2,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7705