Learn More
Sino Biological Transferrin Receptor / TFRC / CD71 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11020R040100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Transferrin Receptor / TFRC / CD71 (rh Transferrin Receptor / TFRC / CD71; Catalog#11020-H07H; AAA61153.1; Cys89-Phe760).
Catalog No. 50-224-8587