Learn More
Sino Biological Recombinant Human Transferrin Receptor/TFR1/CD71 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11020H07H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human TFRC (AAA61153.1) (Cys89-Phe760) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-211-6345