Learn More
Sino Biological Recombinant Human Transferrin Receptor/TFR1/CD71 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11020H07HB100
Encompass_Preferred
A DNA sequence encoding the human TFRC (NP_003225.2) (Cys 89-Phe 760) was expressed with a polyhistidine tag at the N-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6153