Learn More
Sino Biological Transferrin / TF / Serotransferrin Antibody(HRP), Mouse MAb 100ug
Supplier:  Sino Biological 11019MM06H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Transferrin / TF (rh Transferrin; Catalog#11019-H08H; NP_001054.1; Met 1-Pro 698). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-6356