Learn More
ACROBiosystems Recombinant Protein;Mouse Transferrin Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;TRN,Transferrin,TF,DKFZp781D0156,PRO1557,PRO2086
Catalog No. 501657646
Encompass_Preferred
Recombinant Protein;Mouse Transferrin Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Mouse Transferrin, Mouse IgG2a Fc Tag, low endotoxin (TRN-M525b) is expressed from human 293 cells (HEK293). It contains AA Val 20 - His 697 (Accession # Q921I1-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 101.8 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TfR, His Tag (Cat. No. CD1-H5243) at 2 μg/mL (100 μL/well) can bind Mouse Transferrin, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. TRN-M525b) with a linear range of 0.4-12.5 ng/mL (QC tested).
Catalog No. 50-165-7646 Supplier ACROBiosystems Supplier No. TRNM525B1MG
Recombinant Protein;Mouse Transferrin Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Mouse Transferrin, Mouse IgG2a Fc Tag, low endotoxin (TRN-M525b) is expressed from human 293 cells (HEK293). It contains AA Val 20 - His 697 (Accession # Q921I1-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 101.8 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TfR, His Tag (Cat. No. CD1-H5243) at 2 μg/mL (100 μL/well) can bind Mouse Transferrin, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. TRN-M525b) with a linear range of 0.4-12.5 ng/mL (QC tested).