Learn More
ACROBiosystems ACROBiosystems Human Transferrin R / CD71 Protein, His Tag
Catalog No. 501957052
Encompass_Preferred
Recombinant Protein;1MG;Human Transferrin R, His Tag (CD1-H5243) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 77.0 kDa. The protein migrates as 80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Transferrin, His Tag (Cat. No. TRN-H4229) at 2 μg/mL (100 μL/well) can bind Human Transferrin R, His Tag (Cat. No. CD1-H5243) with a linear range of 1-25 ng/mL (QC tested).
Catalog No. 50-195-7052 Supplier ACROBiosystems Supplier No. CD1H52431MG
Recombinant Protein;1MG;Human Transferrin R, His Tag (CD1-H5243) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 77.0 kDa. The protein migrates as 80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Transferrin, His Tag (Cat. No. TRN-H4229) at 2 μg/mL (100 μL/well) can bind Human Transferrin R, His Tag (Cat. No. CD1-H5243) with a linear range of 1-25 ng/mL (QC tested).