Learn More
ACROBiosystems ACROBiosystems Mouse Transferrin R2 Protein, mouse IgG1 Fc Tag, low endotoxin
Supplier:  ACROBiosystems TF2M52691MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse TFR2, Mouse Fc Tag (TF2-M5269) is expressed from human 293 cells (HEK293). It contains AA Arg 103 - Phe 798 (Accession # Q9JKX3-1).;This protein carries a mouse IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 101.1 kDa. The protein migrates as 116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7303