Learn More
ACROBiosystems Recombinant Protein;Cynomolgus Transferrin R / CD71 Protein;HEK293;100UG;TFR,CD71,TFRC,TFR,sTfR,TfR1,Trfr,T9,p90
Catalog No. 501657781
Encompass_Preferred
Recombinant Protein;Cynomolgus Transferrin R / CD71 Protein;100UG;Cynomolgus Transferrin R (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # XP_005545315.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 77.1 kDa. The protein migrates as 75-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Transferrin, His Tag (Cat. No. TRN-H4229) at 5 μg/mL (100 μL/well) can bind Cynomolgus Transferrin R (Cat. No. TFR-C524a) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-165-7781 Supplier ACROBiosystems Supplier No. TFRC524A100UG
Recombinant Protein;Cynomolgus Transferrin R / CD71 Protein;100UG;Cynomolgus Transferrin R (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # XP_005545315.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 77.1 kDa. The protein migrates as 75-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Transferrin, His Tag (Cat. No. TRN-H4229) at 5 μg/mL (100 μL/well) can bind Cynomolgus Transferrin R (Cat. No. TFR-C524a) with a linear range of 0.2-6 ng/mL (QC tested).