Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Transferrin R / CD71, ultra sensitivity (primary amine labeling);HEK293;200UG;TFR,CD71,TFRC,TFR,sTfR,TfR1,Trfr,T9,p90
Catalog No. 501659146
Encompass_Preferred
Recombinant Protein;Biotinylated Human Transferrin R / CD71, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human Transferrin R / CD71 (TFR-H8243) is expressed from human HEK293 cells. It contains AA Cys 89 - Phe 760 (Accession # AAH01188.1). It is the biotinylated form of Human Transferrin R protein, His Tag (Cat # CD1-H5243).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 76 kDa. The protein migrates as 80-85 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human Transferrin, His Tag (Cat. No. TRN-H4229) at 2 g/mL (100 L/well) can bind Biotinylated Human Transferrin R, His Tag (Cat. No. TFR-H8243) with a linear range of 5-156 ng/mL (QC tested).
Catalog No. 50-165-9146 Supplier ACROBiosystems Supplier No. TFRH8243200UG
Recombinant Protein;Biotinylated Human Transferrin R / CD71, ultra sensitivity (primary amine labeling);200UG;MABSol Biotinylated Human Transferrin R / CD71 (TFR-H8243) is expressed from human HEK293 cells. It contains AA Cys 89 - Phe 760 (Accession # AAH01188.1). It is the biotinylated form of Human Transferrin R protein, His Tag (Cat # CD1-H5243).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 76 kDa. The protein migrates as 80-85 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human Transferrin, His Tag (Cat. No. TRN-H4229) at 2 g/mL (100 L/well) can bind Biotinylated Human Transferrin R, His Tag (Cat. No. TFR-H8243) with a linear range of 5-156 ng/mL (QC tested).