Learn More
StatLab Transform FSA Granules
SDP
Catalog No. 22021413
Change view
Click to view available options
Type:
Formalin Absorbent
Catalog No. Type
22-021-413 Formalin Absorbent
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-413 Supplier StatLab Supplier No. CWTFFSA5LB
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
StatLab Transform Foramlin Spill Absorbant (FSA) Granules for large spills
  • Formulated to absorb, deodorize, and neutralize 10% Formalin spills in seconds
  • Just apply to formalin spill and wait one minute
  • 10% Formalin will be absorbed and transformed into small, solid clumps of non-hazardous clay that can be disposed of in your trash receptacle
  • 5 lb. package
Specifications
Type Formalin Absorbent