Learn More
StatLab Transform Spray
SDP
Catalog No. 22021439
Change view
Click to view available options
Type:
Formalin Absorbent
Catalog No. Type
22-021-439 Formalin Absorbent
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-439 Supplier StatLab Supplier No. CWTFSPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab Transform Spray destroys formalin and its fumes in seconds
  • Formulated for personal use in grossing rooms, autopsy rooms, surgical suites, or anywhere else 10% Formalin is used
  • Transform Spray destroys formalin and its fumes in seconds
  • Just spray directly on grossing area, sinks, countertops, or anywhere that formalin fumes are a problem and watch them disappear
Specifications
Type Formalin Absorbent