Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transformation buffer-2, pH 7.5, Thermo Scientific™

Catalog No. AAJ63293EQE
Change view
Click to view available options
Quantity:
125 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63293EQE 125 mL
AAJ63293AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63293EQE Supplier Thermo Scientific Chemicals Supplier No. J63293EQE
Add to Cart

missing translation for 'validMainframeMsg'

For transformation of competent cells

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Transformation buffer-2 is used for transformation of competent cells.

Solubility
Fully miscible in water.

Notes
Contains 10mM MOPS, 75mM calcium chloride, 10mM potassium chloride, and 15% glycerol, pH 7.5
TRUSTED_SUSTAINABILITY

Specifications

pH 7.5
Physical Form Liquid
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.