Learn More
Creative Biomart Transforming Growth Factor-Alpha (TGFA)
Catalog No. 502084293
Encompass_Preferred
TGFA Human Recombinant (40-89) produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 50 amino acids and having a molecular mass of 6kDa. The TGFA is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4293 Supplier Creative Biomart Supplier No. THP0242
TGFA Human Recombinant (40-89) produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 50 amino acids and having a molecular mass of 6kDa. The TGFA is purified by proprietary chromatographic techniques.