Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Transforming Growth Factor-b (TGF-b), Human Natural

Catalog No. MT356039 Shop All Corning Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT356039 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT356039 Supplier Corning™ Supplier No. 356039
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corning™ Transforming Growth Factor-beta (TGF-beta), human natural is a multifunctional protein that plays a central role in the regulation of cell growth and differentiation with either stimulatory or inhibitory effects.

Formulation

Frozen in 36% acetonitrile-0.1% trifluoroacetic acid
Caution: TGF-ß absorbs to glass and plastic surfaces (instructions for dilution are provided with the product)

Source

Human platelets. NOTE: Source material tested for hepatitis B antigen and HIV-1 antibody

Molecular Weight

25 kD

Quality

  • Purity: >95% by SDS-PAGE
  • Tested for ability to inhibit DNA synthesis of Mv 1Lu mink lung cells (ATCC CCL-64)
  • Tested and found negative for bacteria, fungi and mycoplasma
    • Tip

      Recommended concentration: 10-50 pg/mL of medium depending on cell type and application.

      Preparation and Storage

      Stable for at least three months at -70°C (if diluted according to instructions)

      Storage

      Shelf life: ≥ 3 months from date of shipment at -70°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ≥ 3 months from date of shipment (at -70°C)
Purity or Quality Grade >95% by SDS-PAGE
Quantity 5 μg
Source Human Platelets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.