Learn More
STEMCELL Technologies Human Recombinant TGF-beta 1 (Cell Culture Ready), Size: 1 mL
SDP
Supplier:  STEMCELL Technologies 1000856
Encompass
Conveniently and consistently add transforming growth factor beta 1 (TGF-ꞵ1) to your cultures with this cell culture-ready formulation. TGF-β1 binds to serine-threonine kinase type I and II receptors and regulates diverse cellular phenotypes through the SMAD2/3 signal transduction pathway. Use TGF-β1 in combination with basic fibroblast growth factor (bFGF) to support the maintenance of human pluripotent stem cell (hPSC) cultures (Amit et al. Biol Reprod, 2004). TGF-β1 has been used to regulate exogenous cell signaling (Massagué Nat Rev Mol Cell Biol, 2000) and modulate immune cells (Letterio & Roberts Annu Rev Immunol, 1998). Human recombinant TGF-ꞵ1 from STEMCELL is precisely reconstituted to 0.1 mg/ml in 100 mM acetic acid; EC50 is ≤ 1 ng/mL; endotoxin levels verified using the LAL method to ensure consistency for use across multiple applications. Alternative names: Cartilage-inducing factor, Differentiation inhibiting factor, TGF-ꞵ1, TGF-beta-1, Transforming growth factor beta 1.
Catalog No. 50-223-1582
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov