Learn More
MSC Transforming Technologies ESD Gloves, Large, Polyurethane Palm, Dissipative, 12 Pair
Catalog No. 5000766242
Encompass
These anti-static gloves are made from a nylon and conductive carbon blend with a polyurethane-coated palm. They are designed for handling static sensitive items, providing static dissipation and reducing charge generation in clean assembly applications.
  • Coated palms for excellent grip
  • Breathable and comfortable design
  • Made with dissipative nylon to reduce charge generation
  • Resistance of 106–108 per ANSI/ESD SP15.1
  • ESD properties are unaffected by laundering
  • Seamless fine gauge knit for low-linting
  • Washable for repeated use
  • Reduces transfer of fingerprints and oils
  • Finished rolled cuff
  • Ideal for clean assembly applications
Catalog No. 50-007-66242 Supplier MSC Supplier No. 92766344
These anti-static gloves are made from a nylon and conductive carbon blend with a polyurethane-coated palm. They are designed for handling static sensitive items, providing static dissipation and reducing charge generation in clean assembly applications.
  • Coated palms for excellent grip
  • Breathable and comfortable design
  • Made with dissipative nylon to reduce charge generation
  • Resistance of 106–108 per ANSI/ESD SP15.1
  • ESD properties are unaffected by laundering
  • Seamless fine gauge knit for low-linting
  • Washable for repeated use
  • Reduces transfer of fingerprints and oils
  • Finished rolled cuff
  • Ideal for clean assembly applications