Learn More
Electron Microscopy Sciences 12 ml, Transfer Pipettes Straight
SDP
Catalog No. 501899424
Encompass_Preferred
It works as a syringe. Inexpensive and Disposable. Good for liquid transfering tasks. Each pipette has calibration markings from 0 to 12ml in 0.2ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.
Catalog No. 50-189-9424 Supplier Electron Microscopy Sciences Supplier No. 7098600
It works as a syringe. Inexpensive and Disposable. Good for liquid transfering tasks. Each pipette has calibration markings from 0 to 12ml in 0.2ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.