Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Transfer Pipette 7.7 ml, 3.2 ml draw, Sterile individual package
SDP

Catalog No. 501899386
Encompass_Preferred

These pipets are good for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The non-sterile pipets are bulk packed; the sterile are individually packed.
Bulb Draw: 3.2 ml
Drops/mL: 20
OAL: 15.6 cm
Stem diameter: 7.9 cm
Package: Individual

Catalog No. 50-189-9386 Supplier Electron Microscopy Sciences Supplier No. 709601S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These pipets are good for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The non-sterile pipets are bulk packed; the sterile are individually packed.
Bulb Draw: 3.2 ml
Drops/mL: 20
OAL: 15.6 cm
Stem diameter: 7.9 cm
Package: Individual

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.