Learn More
Medicus Health MMassive Transfusion Protocol, MTP Transport Cooler, Blood Bank
SDP
Supplier:  Medicus Health 5307M1
Encompass
Massive Transfusion Protocol (MTP) Blood Bank Transport Cooler combined with standardized components creates a cooler that can easily be reproduced by staff and includes all the tools necessary to comply with AABB and FDA requirements ∙ Massive Transfusion Protocol (MTP) Blood Bank Cooler System includes: (4) Color Coded Ice Packs, (2) Blood Unit Holders, Internal Content Dividers, Document Pocket, Return to Blood Bank Label Smooth surfaces are easy to clean and disinfect with no retention of bio-burden∙ Super duty nylon rope handles (removable)∙ Easy rolling cooler handle extends 39" from the floor ∙ Equipped with 4 inch diameter mag wheels∙ Recessed Zero-Leak Drain Plug for fast drainageRotomolded Coolers ∙ 3" inch thick insulated walls with seal style 15mm gasket ∙ Dual built-In stainless steel security seal loops for maintaining Chain of Custody∙ External Dimensions: 22”W x 18” D x 16”H; Internal Dimensions: 16.5”W x 11.5”D x 14”H
Catalog No. NC3038686