Learn More
Medchemexpress LLC Transglutaminase, Streptoverticillium mobaraense (TG) | 80146-85-6 | 250 MG
SDP
Catalog No. 5000398519
Encompass_Preferred
Transglutaminase, Streptoverticillium mobaraense (TG) is an enzyme that catalyzes the formation of cross-links between protein molecules. It effectively attaches proteins and peptides to various substances including small molecules, polymers, surfaces, and DNA. This enzyme is widely utilized in the food industry for applications in meat, fish, dairy, and baking. It also plays a role in biochemical research, such as in T cell proliferation and HLA class II restriction experiments.
  • Forms cross-links between protein molecules
  • Attaches proteins and peptides to small molecules, polymers, surfaces, and DNA
  • Widely used in food applications
  • Suitable for meat, fish, dairy, and baking industries
  • Solid appearance, white to yellow color
  • Specific activity of ≥10 U/mg protein
  • Supports T cell proliferation experiments
  • Supports HLA class II restriction experiments
Catalog No. 50-003-98519 Supplier Medchemexpress LLC Supplier No. 250MGHYP2962
Transglutaminase, Streptoverticillium mobaraense (TG) is an enzyme that catalyzes the formation of cross-links between protein molecules. It effectively attaches proteins and peptides to various substances including small molecules, polymers, surfaces, and DNA. This enzyme is widely utilized in the food industry for applications in meat, fish, dairy, and baking. It also plays a role in biochemical research, such as in T cell proliferation and HLA class II restriction experiments.
  • Forms cross-links between protein molecules
  • Attaches proteins and peptides to small molecules, polymers, surfaces, and DNA
  • Widely used in food applications
  • Suitable for meat, fish, dairy, and baking industries
  • Solid appearance, white to yellow color
  • Specific activity of ≥10 U/mg protein
  • Supports T cell proliferation experiments
  • Supports HLA class II restriction experiments