Learn More
XpressBio TRANSIL MSA Binding Kit
SDP
Catalog No. NC2333890
Encompass_Preferred
The Sovicell TRANSIL MSA Binding Kit estimates the binding of drugs to mouse serum albumin (MSA) and predicts the plasma protein binding. The assay kit measures the affinity constant (KD) of drugs to albumin and hence allows the calculation of albumin binding even under disease and physiological states that alter the albumin content of human serum. Internal quality controls provide easy assessment of recovery, experiment and data quality.
Catalog No. NC2333890 Supplier XpressBio Supplier No. TPB02302096
Add to Cart
Add to Cart
The Sovicell TRANSIL MSA Binding Kit estimates the binding of drugs to mouse serum albumin (MSA) and predicts the plasma protein binding. The assay kit measures the affinity constant (KD) of drugs to albumin and hence allows the calculation of albumin binding even under disease and physiological states that alter the albumin content of human serum. Internal quality controls provide easy assessment of recovery, experiment and data quality.