Learn More
Thomas Scientific SmartBlue Transilluminator, 115V, US Plug
SDP
Catalog No. 501457741
Encompass
Many new fluorescent dyes have been developed to replace the traditional method of nucleic acid detection using toxic EtBr. The use of these new dyes eliminates the requirement for DNA-damaging UV light, as their fluorescent excitation can be achieved using visible, blue light wavelengths. The Accuris SmartBlue is the perfect transilluminator to use with these new dyes. Blue light, safe for nucleic acids Uniform illumination (UI Technology) Optimal wavelength (465) for green stains Easily removable, 2 position amber shield Optional amber viewing glasses Safer samples, safer for you New fluorophors have been developed to replace the traditional method of nucleic acid detection with toxic Ethidium bromide. They also eliminate the use of DNA damaging UV light, as they are visualized with blue light. The SmartBlue Transilluminator is the perfect partner for these new stains. It emits blue light at a wavelength of 465nm, perfect for excitation of most green fluorophors, including Accuris
Catalog No. 50-145-7741 Supplier Thomas Scientific Supplier No. 1181C30
Many new fluorescent dyes have been developed to replace the traditional method of nucleic acid detection using toxic EtBr. The use of these new dyes eliminates the requirement for DNA-damaging UV light, as their fluorescent excitation can be achieved using visible, blue light wavelengths. The Accuris SmartBlue is the perfect transilluminator to use with these new dyes. Blue light, safe for nucleic acids Uniform illumination (UI Technology) Optimal wavelength (465) for green stains Easily removable, 2 position amber shield Optional amber viewing glasses Safer samples, safer for you New fluorophors have been developed to replace the traditional method of nucleic acid detection with toxic Ethidium bromide. They also eliminate the use of DNA damaging UV light, as they are visualized with blue light. The SmartBlue Transilluminator is the perfect partner for these new stains. It emits blue light at a wavelength of 465nm, perfect for excitation of most green fluorophors, including Accuris