Learn More
Electron Microscopy Sciences EMS SmartBlue Transilluminator
SDP
Catalog No. 501927925
Encompass_Preferred
The SmartBlue™ Transilluminator is the perfect partner for these new stains that are visualized with blue light. It emits blue light at a wavelength of 465 nm, perfect for excitation of most green fluorophors, as well as many protein stains. Exposing DNA to this wavelength does not cause damage and results in a much higher down stream cloning efficiency as compared to even short UV light exposures. In addition, the blue light is not damaging to the skin and eyes.
An array of super bright LEDs with a long, 30,000 hour service life provide the light source for the SmartBlue™ Transilluminator. Unlike a UV Transilluminator, the filters in the SmartBlue™ will not solarize and degrade in performance over time. Gel bands can be excised directly on the scratch resistant, glass viewing surface.
Catalog No. 50-192-7925 Supplier Electron Microscopy Sciences Supplier No. 8705801
The SmartBlue™ Transilluminator is the perfect partner for these new stains that are visualized with blue light. It emits blue light at a wavelength of 465 nm, perfect for excitation of most green fluorophors, as well as many protein stains. Exposing DNA to this wavelength does not cause damage and results in a much higher down stream cloning efficiency as compared to even short UV light exposures. In addition, the blue light is not damaging to the skin and eyes.
An array of super bright LEDs with a long, 30,000 hour service life provide the light source for the SmartBlue™ Transilluminator. Unlike a UV Transilluminator, the filters in the SmartBlue™ will not solarize and degrade in performance over time. Gel bands can be excised directly on the scratch resistant, glass viewing surface.