Learn More
COLDER PRODUCTS COMPANY TRANSITION 3/4 FEMALE 25/PKTR
Supplier:  COLDER PRODUCTS COMPANY MPX3301239
Encompass_Preferred
CPC MPX Series Connectors and Configurations Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber For Use With 12 7mm Polysulfone Overall Length 1 4 in 36 mm Sanitary Coupling Body Height 1 in 25 mm Maximum Pressure 60 psi
Catalog No. 50-313-1414