Learn More
Epigentek Inc Recombinant Transmembrane Protease Serine 2 Protein, Partial (20 µg)
SDP
Supplier:  Epigentek Inc E800161
Encompass_Preferred
The recombinant Transmembrane protease serine 2 (TMPRSS2) is produced by the expression of a target DNA sequence with 6xHis, N-terminal tag(s), in the yeast expression system. The target DNA sequence encodes the 106-492aa region of the TMPRSS2. This protein predicts a molecular weight of 44.8 kDa. Its purity is greater than 90% as determined by SDS-PAGE. And the bio-activity test of this protein is ongoing.
Catalog No. 50-201-9363