Learn More
Perkin Elmer US LLC Transmission Holder for LAMBDA 365
Catalog No. 502105508
Encompass_Preferred
PerkinElmer offers a variety of advanced transmission holders. These holders support transmission measurements on solid samples (films, filters, glass, plastics, etc.). The springloaded clamps securely hold objects of varying thicknesses.
Catalog No. 50-210-5508 Supplier Perkin Elmer US LLC Supplier No. N4101019
PerkinElmer offers a variety of advanced transmission holders. These holders support transmission measurements on solid samples (films, filters, glass, plastics, etc.). The springloaded clamps securely hold objects of varying thicknesses.