Learn More
Leica Biosystems BF Transmitted Light Condenser for Leica DM-Series Microscopes and Compatible Motorized Scanning Stages
Catalog No. NC3365974
Encompass_Preferred
Bright-field transmitted light condenser designed for Leica Microsystems modular microscope systems. It provides bright-field illumination for transmitted light microscopy and integrates with DM-series bases and motorized scanning stages to support routine bright-field imaging of slides in research and pathology workflows.
  • Provides bright-field transmitted light illumination for routine microscopy
  • Integrates with modular microscope bases and motorized scanning stages
  • Optimized for transmitted light bright-field imaging applications
  • BF-only configuration prevents use of DIC/phase condensers when paired with BF-only stage adapters
  • Intended for research and clinical pathology use where bright-field contrast is required
Catalog No. NC3365974 Supplier Leica Biosystems Supplier No. 11505321
Bright-field transmitted light condenser designed for Leica Microsystems modular microscope systems. It provides bright-field illumination for transmitted light microscopy and integrates with DM-series bases and motorized scanning stages to support routine bright-field imaging of slides in research and pathology workflows.
  • Provides bright-field transmitted light illumination for routine microscopy
  • Integrates with modular microscope bases and motorized scanning stages
  • Optimized for transmitted light bright-field imaging applications
  • BF-only configuration prevents use of DIC/phase condensers when paired with BF-only stage adapters
  • Intended for research and clinical pathology use where bright-field contrast is required