Learn More
Cole-Parmer Ashcroft IXLDP5IWC Low Diff Pressure Transmitter, 5.0" WC, 4-20mA, 0.25%
Catalog No. 50992718 Shop All Cole-Parmer Products
Encompass_Preferred
Designed for the measurement and control of very low pressure and flow in industrial and process plant environments. The transmitter features s rugged NEMA4X enclosure, built-in electrical terminal box isolated from the electronics and threaded process connections.
Catalog No. 50-992-718 Supplier Cole-Parmer Supplier No. 6843931
Designed for the measurement and control of very low pressure and flow in industrial and process plant environments. The transmitter features s rugged NEMA4X enclosure, built-in electrical terminal box isolated from the electronics and threaded process connections.